siRNA Details
Gene Symbol: RGD1306063 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106026.2 | NP_001099496.1 | 4 | 298 | ||
XM_006251730.2 | XP_006251792.1 | 4 | 332 | |||
XM_006251731.2 | XP_006251793.1 | 4 | 329 | |||
XM_006251732.2 | XP_006251794.1 | 4 | 350 | |||
XM_006251733.2 | XP_006251795.1 | 4 | 334 | |||
XM_006251736.2 | XP_006251798.1 | 4 | 332 | |||
GenBank mRNA | BC168755.1 | - | 4 | 298 | 6887151 |
