siRNA Details
Gene Symbol: RGD1564324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106035.1 | NP_001099505.1 | 1 | 63 | ||
XM_008770665.1 | XP_008768887.1 | 2 | 304 | |||
XM_008770666.1 | XP_008768888.1 | 2 | 304 | |||
XM_008770667.1 | XP_008768889.1 | 1 | 126 | |||
XM_008770668.1 | XP_008768890.1 | 2 | 304 | |||
XM_008770669.1 | XP_008768891.1 | 2 | 304 | |||
XM_008770670.1 | XP_008768892.1 | 2 | 304 | |||
XM_008770671.1 | XP_008768893.1 | 2 | 304 |
