siRNA Details
Gene Symbol: Olfm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106052.1 | NP_001099522.1 | 5 | 1455 | ||
XM_008770863.1 | XP_008769085.1 | 5 | 2282 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106052.1 | NP_001099522.1 | 5 | 1455 | ||
XM_008770863.1 | XP_008769085.1 | 5 | 2282 |