siRNA Details
Gene Symbol: Slitrk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106057.1 | NP_001099527.1 | 2 | 2929 | ||
XM_006252443.2 | XP_006252505.1 | 2 | 3162 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slitrk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106057.1 | NP_001099527.1 | 2 | 2929 | ||
XM_006252443.2 | XP_006252505.1 | 2 | 3162 |