siRNA Details
Gene Symbol: B3gnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106068.1 | NP_001099538.1 | 2 | 410 | ||
XM_008771081.1 | XP_008769303.1 | 1 | 360 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: B3gnt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106068.1 | NP_001099538.1 | 2 | 410 | ||
XM_008771081.1 | XP_008769303.1 | 1 | 360 |