siRNA Details
Gene Symbol: LOC100910087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751594.3 | XP_003751642.1 | 581 | |||
XM_224829.5 | XP_224829.3 | 581 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100910087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751594.3 | XP_003751642.1 | 581 | |||
XM_224829.5 | XP_224829.3 | 581 |