siRNA Details
Gene Symbol: RGD1564788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253254.2 | XP_006253316.1 | 6 | 1414 | ||
XM_006253256.2 | XP_006253318.1 | 6 | 843 | |||
XM_008771311.1 | XP_008769533.1 | 6 | 1414 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001122822, NM_011721, BC050921, AF091215, AF241636, and others.
Gene Ontology Categories:
