siRNA Details
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014772.1 | NP_001014772.1 | 2 | 145 | ||
GenBank mRNA | AY953138.1 | - | 2 | 145 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014772.1 | NP_001014772.1 | 2 | 145 | ||
GenBank mRNA | AY953138.1 | - | 2 | 145 |