siRNA Details
Gene Symbol: Mtrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039003.1 | NP_001034092.1 | 5, 6 | 551 | ||
XM_006227813.2 | XP_006227875.1 | 7, 8 | 1180 | |||
XM_006227814.1 | XP_006227876.1 | 5, 6 | 844 | |||
XM_008758680.1 | XP_008756902.1 | 4, 5 | 1254 | |||
GenBank mRNA | BC100107.1 | - | 5, 6 | 551 | 7379770 |
