siRNA Details
Gene Symbol: Lman2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115024.2 | NP_001108496.1 | 1 | 168 | ||
GenBank mRNA | BC158808.1 | - | 1 | 168 | 7324877 | |
FQ215124.1 | - | 1 | 349 | |||
FQ227127.1 | - | 1 | 52 | |||
FQ230263.1 | - | 1 | 166 |
