siRNA Details
Gene Symbol: Faf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017445.1 | NP_001017445.1 | 8 | 624 | ||
XM_006253617.2 | XP_006253679.1 | 7 | 649 | |||
XM_008771488.1 | XP_008769710.1 | 5 | 433 | |||
GenBank mRNA | BC079356.1 | - | 7 | 623 | ||
BC091224.1 | - | 8 | 624 | 7128668 |
