siRNA Details
Gene Symbol: Nrsn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106109.2 | NP_001099579.2 | 4 | 426 | ||
GenBank mRNA | BC166881.1 | - | 4 | 426 | 6891333 | |
BC166919.1 | - | 4 | 426 | 6919003 |
