siRNA Details
Gene Symbol: Usp6nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106120.1 | NP_001099590.1 | 15 | 2081 | ||
XM_006254232.2 | XP_006254294.1 | 14 | 2325 | |||
XM_006254233.1 | XP_006254295.1 | 14 | 1950 | |||
XM_006254236.2 | XP_006254298.1 | 12 | 3431 |
