siRNA Details
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106140.1 | NP_001099610.1 | 25 | 3332 | ||
XM_006254842.2 | XP_006254904.1 | 25 | 3355 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Atp8b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106140.1 | NP_001099610.1 | 25 | 3332 | ||
XM_006254842.2 | XP_006254904.1 | 25 | 3355 |