siRNA Details
Gene Symbol: RGD1562699 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106141.1 | NP_001099611.1 | 2 | 166 | ||
XM_006254846.2 | XP_006254908.1 | 5 | 967 | |||
XM_006254847.2 | XP_006254909.1 | 5 | 454 | |||
XM_006254848.2 | XP_006254910.1 | 5 | 472 | |||
XM_008772119.1 | XP_008770341.1 | 5 | 398 | |||
XM_008772120.1 | XP_008770342.1 | 5 | 385 | |||
XM_008772122.1 | XP_008770344.1 | 5 | 488 | |||
XM_008772123.1 | XP_008770345.1 | 5 | 942 |
