siRNA Details
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271074.1 | NP_001258003.1 | 4, 5 | 211 | ||
NM_001271077.1 | NP_001258006.1 | 4, 5 | 211 | |||
NM_019154.1 | NP_062027.1 | 4, 5 | 202 | |||
GenBank mRNA | U01245.2 | - | 3, 4 | 170 | ||
U67130.1 | - | 4, 5 | 202 |

More Information
Other Species Targeted

Gene Ontology Categories:
