siRNA Details
Gene Symbol: Amelx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271073.1 | NP_001258002.1 | 5 | 322 | ||
NM_001271074.1 | NP_001258003.1 | 5 | 394 | |||
NM_001271077.1 | NP_001258006.1 | 5 | 394 | |||
NM_019154.1 | NP_062027.1 | 5 | 385 | |||
GenBank mRNA | U01245.2 | - | 4 | 353 | ||
U51195.1 | - | 5 | 395 | |||
U60562.1 | - | 5 | 323 | |||
U67130.1 | - | 5 | 385 |

More Information
Other Species Targeted

Gene Ontology Categories:
