siRNA Details
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100945.1 | NP_001094415.1 | 5, 6 | 1101 | ||
GenBank mRNA | AY724539.1 | - | 4, 5 | 744 | ||
BC129112.1 | - | 5, 6 | 1101 | 7112017 |

More Information
Other Species Targeted

Gene Ontology Categories:
