siRNA Details
Gene Symbol: Nudt21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039004.1 | NP_001034093.1 | 5, 6 | 623 | ||
XM_006255132.2 | XP_006255194.1 | 5, 6 | 1256 | |||
GenBank mRNA | BC098748.1 | - | 5, 6 | 623 | 7365565 | |
BC168149.1 | - | 5, 6 | 1262 | 7457021 | ||
FQ213930.1 | - | 5, 6 | 666 | |||
FQ229498.1 | - | 5, 6 | 664 |

More Information
Other Species Targeted

Gene Ontology Categories:
