siRNA Details
Gene Symbol: Tox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106171.1 | NP_001099641.1 | 2 | 269 | ||
XM_006255187.2 | XP_006255249.1 | 3 | 418 | |||
XM_006255189.2 | XP_006255251.1 | 3 | 415 |
