siRNA Details
Gene Symbol: Ces4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106176.1 | NP_001099646.1 | 6, 7 | 957 | ||
XM_006255449.2 | XP_006255511.1 | 6, 7 | 995 | |||
XM_006255451.1 | XP_006255513.1 | 5, 6 | 543 | |||
XM_008772476.1 | XP_008770698.1 | 5, 6 | 612 | |||
XM_008772477.1 | XP_008770699.1 | 6, 7 | 995 | |||
GenBank mRNA | FQ220291.1 | - | 6, 7 | 800 |
