siRNA Details
Gene Symbol: Sf3b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106187.2 | NP_001099657.2 | 19 | 2765 | ||
XM_006255620.2 | XP_006255682.1 | 19 | 2900 | |||
GenBank mRNA | BC168883.1 | - | 13 | 1670 |
