siRNA Details
Gene Symbol: Casq2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017131.2 | NP_058827.3 | 4 | 647 | ||
GenBank mRNA | BC072547.1 | - | 4 | 634 | 7105911 | |
CK602439.1 | - | 4 | 647 | |||
U33287.1 | - | 4 | 628 |
