siRNA Details
Gene Symbol: Epha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031564.1 | NP_113752.1 | 5 | 1104 | ||
XM_006247976.2 | XP_006248038.1 | 5 | 1188 | |||
XM_006247977.2 | XP_006248039.1 | 5 | 2593 | |||
XM_006247978.2 | XP_006248040.1 | 4 | 1117 | |||
XM_008768529.1 | XP_008766751.1 | 4 | 2227 | |||
XM_008768530.1 | XP_008766752.1 | 4 | 991 | |||
GenBank mRNA | U69278.1 | - | 5 | 1104 |
