siRNA Details
Gene Symbol: Snca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019169.2 | NP_062042.1 | 2, 3 | 279 | ||
XM_006236591.2 | XP_006236653.1 | 2, 3 | 560 | |||
GenBank mRNA | AF007758.1 | - | 1, 2 | 142 | ||
AY550005.1 | - | 1, 2 | 142 | |||
AY550006.1 | - | 1, 2 | 142 | |||
BC087682.1 | - | 2, 3 | 279 | 7321273 | ||
FQ212580.1 | - | 2, 3 | 256 | |||
FQ227096.1 | - | 2, 3 | 270 | |||
S73007.1 | - | 2, 3 | 210 | |||
S73008.1 | - | 2, 3 | 210 |
