siRNA Details
Gene Symbol: Aurkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106221.1 | NP_001099691.1 | 2 | 215 | ||
XM_008758900.1 | XP_008757122.1 | 5 | 579 | |||
XM_008758901.1 | XP_008757123.1 | 6 | 682 |
