siRNA Details
Gene Symbol: Ptgir | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077644.1 | NP_001071112.1 | 1 | 874 | ||
GenBank mRNA | D28966.1 | - | 1 | 874 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
