siRNA Details
Gene Symbol: Fam98c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198584.1 | NP_001185513.1 | 7, 8 | 912 | ||
XM_006228717.2 | XP_006228779.1 | 7, 8 | 942 | |||
GenBank mRNA | BC079080.1 | - | 6, 7 | 950 | 7122702 | |
CO564588.1 | - | 5, 6 | 689 |
