siRNA Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100831.1 | NP_001094301.1 | 1 | 322 | ||
GenBank mRNA | BC105894.1 | - | 1 | 87 | ||
CD371338.1 | - | 1 | 332 | |||
FQ213223.1 | - | 1 | 122 | |||
FQ214314.1 | - | 1 | 119 | |||
FQ223219.1 | - | 1 | 165 | |||
FQ233693.1 | - | 1 | 153 |
