siRNA Details
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100831.1 | NP_001094301.1 | 2 | 451 | ||
GenBank mRNA | BC105894.1 | - | 2 | 216 | ||
CD371338.1 | - | 2 | 461 | |||
FQ213223.1 | - | 2 | 251 | |||
FQ214314.1 | - | 2 | 248 | |||
FQ223219.1 | - | 2 | 294 | |||
FQ233693.1 | - | 2 | 282 |
