siRNA Details
Gene Symbol: Ffar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005877.1 | NP_001005877.1 | 1 | 595 | ||
GenBank mRNA | AB106675.1 | - | 1 | 595 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ffar2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005877.1 | NP_001005877.1 | 1 | 595 | ||
GenBank mRNA | AB106675.1 | - | 1 | 595 |
Set Membership: |
![]() |