siRNA Details
Gene Symbol: Akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106259.3 | NP_001099729.2 | 5 | 773 | ||
XM_006229003.2 | XP_006229065.1 | 5 | 1249 | |||
XM_006229004.2 | XP_006229066.1 | 5 | 949 | |||
XM_006229005.2 | XP_006229067.1 | 5 | 925 | |||
XM_008759335.1 | XP_008757557.1 | 4 | 502 | |||
GenBank mRNA | EV773331.1 | - | 4 | 689 |
