siRNA Details
Gene Symbol: RGD1305713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024749.1 | NP_001019920.1 | 4 | 442 | ||
XM_006229429.2 | XP_006229491.1 | 4 | 2147 | |||
GenBank mRNA | BC094529.1 | - | 4 | 442 | 7320095 |
Gene Symbol: LOC100363116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750235.3 | XP_003750283.1 | 1 | 711 |
