siRNA Details
Gene Symbol: Crebzf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271791.1 | NP_001258720.1 | 1 | 1030 | ||
XM_006229688.2 | XP_006229750.1 | 1 | 1395 | |||
XM_008759646.1 | XP_008757868.1 | 1 | 1395 | |||
GenBank mRNA | FN797334.1 | - | 1 | 79 |
