siRNA Details
Gene Symbol: Fam160a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005538.2 | NP_001005538.2 | 8 | 1636 | ||
XM_006229931.2 | XP_006229993.1 | 8 | 1670 | |||
XM_006229932.2 | XP_006229994.1 | 8 | 1669 | |||
XM_008759708.1 | XP_008757930.1 | 8 | 1670 | |||
GenBank mRNA | BC082011.1 | - | 8 | 1622 | 7132039 |
