siRNA Details
Gene Symbol: Olr229 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000204.1 | NP_001000204.1 | 804 |
Gene Symbol: LOC100911839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006229964.2 | XP_006230026.1 | 1 | 947 |
