siRNA Details
Gene Symbol: Palb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191603.1 | NP_001178532.1 | 7 | 2418 | ||
XM_008759711.1 | XP_008757933.1 | 5 | 3594 | |||
XM_008759712.1 | XP_008757934.1 | 6 | 2502 | |||
GenBank mRNA | FQ211178.1 | - | 7 | 2567 |
