siRNA Details
Gene Symbol: Prkacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077645.1 | NP_001071113.1 | 3, 4 | 296 | ||
XM_006233451.2 | XP_006233513.1 | 3, 4 | 1013 | |||
XM_006233452.2 | XP_006233514.1 | 3, 4 | 2518 | |||
XM_006233453.2 | XP_006233515.1 | 3, 4 | 760 | |||
GenBank mRNA | D10770.1 | - | 3, 4 | 296 | ||
X53261.1 | - | 2, 3 | 191 |
