siRNA Details
Gene Symbol: Gprc5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106304.1 | NP_001099774.1 | 2 | 352 | ||
XM_006230109.2 | XP_006230171.1 | 2 | 326 | |||
XM_006230110.2 | XP_006230172.1 | 2 | 384 | |||
XM_008759715.1 | XP_008757937.1 | 2 | 399 | |||
GenBank mRNA | BC166986.1 | - | 2 | 316 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
