siRNA Details
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106316.1 | NP_001099786.1 | 9 | 932 | ||
XM_006230532.2 | XP_006230594.1 | 9 | 1748 | |||
GenBank mRNA | BC166496.1 | - | 877 | 7111527 |
