siRNA Details
Gene Symbol: Chid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047854.2 | NP_001041319.2 | 10, 11 | 1031 | ||
GenBank mRNA | BC127546.1 | - | 10, 11 | 1031 | 7324297 | |
FQ228688.1 | - | 11, 12 | 1092 |
Gene Symbol: LOC100911881 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760034.1 | XP_008758256.1 | 1105 | |||
XM_008760035.1 | XP_008758257.1 | 1127 |
