siRNA Details
Gene Symbol: Cars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106319.1 | NP_001099789.1 | 17 | 1832 | ||
XM_006230713.2 | XP_006230775.1 | 18 | 2088 | |||
GenBank mRNA | BC166707.1 | - | 17 | 1832 |
