siRNA Details
Gene Symbol: Efemp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005907.2 | NP_001005907.1 | 4 | 410 | ||
NM_001277341.1 | NP_001264270.1 | 4 | 410 | |||
XM_006230742.2 | XP_006230804.1 | 4 | 750 | |||
XM_006230743.1 | XP_006230805.1 | 4 | 401 | |||
XM_006230744.2 | XP_006230806.1 | 4 | 407 | |||
GenBank mRNA | BC083804.1 | - | 4 | 348 | 7109182 | |
FM037652.1 | - | 4 | 372 |
