siRNA Details
Gene Symbol: B3gat3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128184.1 | NP_001121656.1 | 4, 5 | 1078 | ||
GenBank mRNA | BC166510.1 | - | 4, 5 | 956 | 7127430 | |
BC168710.1 | - | 4, 5 | 1059 | 8364370 |
