siRNA Details
Gene Symbol: Pdzd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135836.1 | NP_001129308.1 | 8 | 1543 | ||
XM_006229562.2 | XP_006229624.1 | 8 | 2102 | |||
XM_006229563.2 | XP_006229625.1 | 8 | 1953 | |||
XM_006229564.2 | XP_006229626.1 | 7 | 2039 | |||
XM_006229565.2 | XP_006229627.1 | 6 | 1757 |
