siRNA Details
Gene Symbol: Plgrkt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106347.1 | NP_001099817.1 | 6 | 726 | ||
XM_006231203.2 | XP_006231265.1 | 7 | 787 | |||
XM_006231204.2 | XP_006231266.1 | 7 | 691 | |||
XM_006231205.2 | XP_006231267.1 | 6 | 630 | |||
XM_008760323.1 | XP_008758545.1 | 4 | 816 |
