siRNA Details
Gene Symbol: Tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017185.1 | NP_058881.1 | 4, 5 | 225 | ||
GenBank mRNA | FQ216337.1 | - | 4, 5 | 267 | ||
FQ217026.1 | - | 5, 6 | 267 | |||
FQ217454.1 | - | 5, 6 | 275 | |||
FQ217985.1 | - | 5, 6 | 267 | |||
FQ218058.1 | - | 4, 5 | 299 | |||
FQ223581.1 | - | 5, 6 | 267 | |||
FQ223772.1 | - | 5, 6 | 273 | |||
FQ224443.1 | - | 5, 6 | 268 | |||
FQ224464.1 | - | 5, 6 | 267 | |||
FQ224489.1 | - | 5, 6 | 267 | |||
M73701.1 | - | 4, 5 | 225 |
