siRNA Details
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053304.1 | NP_445756.1 | 2 | 251 | ||
GenBank mRNA | BC133728.1 | - | 2 | 235 | 7125649 | |
CB693258.1 | - | 2 | 259 | |||
Z78279.1 | - | 2 | 140 |
