siRNA Details
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106362.2 | NP_001099832.2 | 16 | 3090 | ||
XM_006231449.2 | XP_006231511.1 | 17 | 3696 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106362.2 | NP_001099832.2 | 16 | 3090 | ||
XM_006231449.2 | XP_006231511.1 | 17 | 3696 |