siRNA Details
Gene Symbol: Papss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106375.2 | NP_001099845.2 | 8 | 967 | ||
XM_006231274.2 | XP_006231336.1 | 9 | 1127 | |||
GenBank mRNA | BC169061.1 | - | 8 | 967 | 5621354 |
